Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-4'-fluorobutyrophenone, 96%

$37.36 - $388.74

Chemical Identifiers

Chemical Name or Material 4-Chloro-4'-fluorobutyrophenone
CAS 3874-54-2
Molecular Formula C10H10ClFO
Formula Weight 200.64
MDL Number MFCD00001007
Beilstein 608741
Synonym 3-Chloropropyl 4-fluorophenyl ketone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1112114

  SDS
alfa aesar™
A1112114
25g Each for $37.36

AAA1112122

  SDS
alfa aesar™
A1112122
100g Each for $102.77

AAA1112136

  SDS
alfa aesar™
A1112136
500g Each for $388.74
Description & Specifications

Specifications

Formula Weight 200.64
Chemical Name or Material 4-Chloro-4'-fluorobutyrophenone
TSCA TSCA
Assay 96%
Molecular Formula C10H10ClFO
MDL Number MFCD00001007
CAS 3874-54-2
Beilstein 608741
Synonym 3-Chloropropyl 4-fluorophenyl ketone
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302
Harmful if swallowed.
Health Hazard 3 GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2°C to 8°C
Flash Point 152°C (306°F)
Boiling Point 120° to 122°C (0.7mmHg)
Melting Point 5° to 6°C
Quantity 25g
Safety and Handling

Warning

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302
Harmful if swallowed.

2°C to 8°C