missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-4'-fluorobutyrophenone, 96%

Manufacturer:  Alfa Aesar™ A1112136

 View more versions of this product

Catalog No. AAA1112136



Specifications

Specifications

4-Chloro-4'-fluorobutyrophenone
120°C to 122°C (0.7mmHg)
500g
96%
MFCD00001007
3-Chloropropyl 4-fluorophenyl ketone
C1=CC(=CC=C1C(=O)CCCCl)F
200.637
200.64
152°C (306°F)
5°C to 6°C
3874-54-2
C10H10ClFO
608741
HXAOUYGZEOZTJO-UHFFFAOYSA-N
4-chloro-1-(4-fluorophenyl)butan-1-one
19750

Chemical Identifiers

3874-54-2
200.637
HXAOUYGZEOZTJO-UHFFFAOYSA-N
19750
C1=CC(=CC=C1C(=O)CCCCl)F
C10H10ClFO
MFCD00001007
3-Chloropropyl 4-fluorophenyl ketone
4-chloro-1-(4-fluorophenyl)butan-1-one
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Recommended Storage : 2°C to 8°C

SDS Documents