Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-3-(trifluoromethyl)phenylacetic acid, 97%

$81.43 - $243.20

Chemical Identifiers

Chemical Name or Material 4-Chloro-3-(trifluoromethyl)phenylacetic acid
CAS 22902-86-9
Molecular Formula C9H6ClF3O2
Formula Weight 238.59
MDL Number MFCD04116012
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2629503

 
alfa aesar™
H2629503
1g Each for $243.20

AAH26295MD

 
alfa aesar™
H26295MD
250mg Each for $81.43
Description & Specifications

Specifications

Formula Weight 238.59
Chemical Name or Material 4-Chloro-3-(trifluoromethyl)phenylacetic acid
Odor Odorless
Assay 97%
Molecular Formula C9H6ClF3O2
MDL Number MFCD04116012
CAS 22902-86-9
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 82° to 84°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.