Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-3-(trifluoromethyl)phenyl isocyanate, 97%

Chemical Identifiers

Chemical Name or Material 4-Chloro-3-(trifluoromethyl)phenyl isocyanate
CAS 327-78-6
Molecular Formula C8H3ClF3NO
Formula Weight 221.57
MDL Number MFCD00013874
UN Number UN2811
Beilstein 522032
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1201603

  SDS
alfa aesar™
L1201603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1201606

  SDS
alfa aesar™
L1201606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 221.57
MDL Number MFCD00013874
CAS 327-78-6
Chemical Name or Material 4-Chloro-3-(trifluoromethyl)phenyl isocyanate
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flash Point 99°C (210°F)
Molecular Formula C8H3ClF3NO
UN Number UN2811
Assay 97%
Odor Pungent
Health Hazard 3 GHS P Statement
P284a-P261-P280-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 86° to 90°C (14mmHg)
Quantity 1g
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Melting Point 34° to 36°C
Recommended Storage 2° to 8°C
Beilstein 522032
Safety and Handling

Danger

GHS P Statement
P284a-P261-P280-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C