Learn More
4-Chloro-3-(trifluoromethyl)phenyl isocyanate, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1201606
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-Chloro-3-(trifluoromethyl)phenyl isocyanate | |
34°C to 36°C | |
99°C (210°F) | |
C8H3ClF3NO | |
5 g | |
522032 | |
4-chloro-3-trifluoromethyl phenyl isocyanate, 1-chloro-4-isocyanato-2-trifluoromethyl benzene, 4-chloro-3-trifluoromethyl phenylisocyanate, benzene, 1-chloro-4-isocyanato-2-trifluoromethyl, 4-chloro-3-trifluoromethylphenyl isocyanate, 4-chloro-3-trifluoromethoxy phenyl isocyanate, 1-chloro-4-isocyanato-2-trifluoromethyl-benzene, 4-chloro-3-trifluoromethyl benzenisocyanate, 3-trifluoromethyl-4-chloro-phenyl isocyanate, pubchem5027 | |
C1=CC(=C(C=C1N=C=O)C(F)(F)F)Cl | |
221.563 | |
221.57 |
327-78-6 | |
86°C to 90°C (14 mmHg) | |
Pungent | |
MFCD00013874 | |
UN2811 | |
Moisture sensitive | |
NBJZEUQTGLSUOB-UHFFFAOYSA-N | |
1-chloro-4-isocyanato-2-(trifluoromethyl)benzene | |
2733265 | |
97% |
Chemical Identifiers
327-78-6 | |
221.563 | |
NBJZEUQTGLSUOB-UHFFFAOYSA-N | |
2733265 | |
C1=CC(=C(C=C1N=C=O)C(F)(F)F)Cl |
C8H3ClF3NO | |
MFCD00013874 | |
4-chloro-3-trifluoromethyl phenyl isocyanate, 1-chloro-4-isocyanato-2-trifluoromethyl benzene, 4-chloro-3-trifluoromethyl phenylisocyanate, benzene, 1-chloro-4-isocyanato-2-trifluoromethyl, 4-chloro-3-trifluoromethylphenyl isocyanate, 4-chloro-3-trifluoromethoxy phenyl isocyanate, 1-chloro-4-isocyanato-2-trifluoromethyl-benzene, 4-chloro-3-trifluoromethyl benzenisocyanate, 3-trifluoromethyl-4-chloro-phenyl isocyanate, pubchem5027 | |
1-chloro-4-isocyanato-2-(trifluoromethyl)benzene |
Safety and Handling
GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P260-P264b-P270-P271-P272-P280-P284-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P330-P333+P313-P362-P501c
H302-H315-H317-H318-H330
DOTInformation : Hazard Class: 6.1; Packaging Group: I
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only