Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-3-(trifluoromethyl)phenol, 97%

Chemical Identifiers

Chemical Name or Material 4-Chloro-3-(trifluoromethyl)phenol
CAS 6294-93-5
Molecular Formula C7H4ClF3O
Formula Weight 196.56
MDL Number MFCD00042525
UN Number UN3261
Beilstein 2261083
Synonym 2-Chloro-5-hydroxybenzotrifluoride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2058003

  SDS
alfa aesar™
B2058003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2058006

  SDS
alfa aesar™
B2058006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2058014

  SDS
alfa aesar™
B2058014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 196.56
Chemical Name or Material 4-Chloro-3-(trifluoromethyl)phenol
Odor Phenol-like
Assay 97%
Molecular Formula C7H4ClF3O
MDL Number MFCD00042525
CAS 6294-93-5
UN Number UN3261
Beilstein 2261083
Synonym 2-Chloro-5-hydroxybenzotrifluoride
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318-H302-H312
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
RTECS Number SK6040000
Boiling Point 120° to 122°C (27mmHg)
Melting Point 42° to 46°C
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318-H302-H312
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.