Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-3-(trifluoromethyl)benzonitrile, 97%

Chemical Identifiers

Chemical Name or Material 4-Chloro-3-(trifluoromethyl)benzonitrile
CAS 1735-54-2
Molecular Formula C8H3ClF3N
Formula Weight 205.57
MDL Number MFCD00190117
UN Number UN3439
Beilstein 2103768
Synonym 2-Chloro-5-cyanobenzotrifluoride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2011903

  SDS
alfa aesar™
B2011903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 205.57
Chemical Name or Material 4-Chloro-3-(trifluoromethyl)benzonitrile
Odor Characteristic
Assay 97%
Molecular Formula C8H3ClF3N
MDL Number MFCD00190117
CAS 1735-54-2
UN Number UN3439
Beilstein 2103768
Synonym 2-Chloro-5-cyanobenzotrifluoride
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
Boiling Point 100° to 105°C (11mmHg)
Melting Point 64° to 68°C
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.