missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Chloro-3-(trifluoromethyl)benzoic Acid 98.0+%, TCI America™
$26.54 - $92.90
Chemical Identifiers
CAS | 1737-36-6 |
---|---|
Molecular Formula | C8H3ClF3O2 |
Molecular Weight (g/mol) | 223.56 |
MDL Number | MFCD01631468 |
InChI Key | PPHHAZOVVZBSCM-UHFFFAOYSA-M |
Synonym | 4-chloro-3-trifluoromethyl benzoic acid, benzoic acid, 4-chloro-3-trifluoromethyl, 5-carboxy-2-chlorobenzotrifluoride, 4-chloro-3-trifluoromethyl-benzoic acid, 3-trifluoromethyl-4-chlorobenzoic acid, pubchem4588, intermediates-zcf02292, ksc496a8t, 4-chloro-3-trifluoromethyl |
PubChem CID | 2773857 |
IUPAC Name | 4-chloro-3-(trifluoromethyl)benzoate |
SMILES | [O-]C(=O)C1=CC=C(Cl)C(=C1)C(F)(F)F |
Chemical Identifiers
1737-36-6 | |
223.56 | |
PPHHAZOVVZBSCM-UHFFFAOYSA-M | |
2773857 | |
[O-]C(=O)C1=CC=C(Cl)C(=C1)C(F)(F)F |
C8H3ClF3O2 | |
MFCD01631468 | |
4-chloro-3-trifluoromethyl benzoic acid, benzoic acid, 4-chloro-3-trifluoromethyl, 5-carboxy-2-chlorobenzotrifluoride, 4-chloro-3-trifluoromethyl-benzoic acid, 3-trifluoromethyl-4-chlorobenzoic acid, pubchem4588, intermediates-zcf02292, ksc496a8t, 4-chloro-3-trifluoromethyl | |
4-chloro-3-(trifluoromethyl)benzoate |
Specifications
1737-36-6 | |
White | |
MFCD01631468 | |
4-chloro-3-trifluoromethyl benzoic acid, benzoic acid, 4-chloro-3-trifluoromethyl, 5-carboxy-2-chlorobenzotrifluoride, 4-chloro-3-trifluoromethyl-benzoic acid, 3-trifluoromethyl-4-chlorobenzoic acid, pubchem4588, intermediates-zcf02292, ksc496a8t, 4-chloro-3-trifluoromethyl | |
[O-]C(=O)C1=CC=C(Cl)C(=C1)C(F)(F)F | |
223.56 | |
224.56 | |
Crystalline Powder |
160°C | |
C8H3ClF3O2 | |
1 g | |
PPHHAZOVVZBSCM-UHFFFAOYSA-M | |
4-chloro-3-(trifluoromethyl)benzoate | |
2773857 | |
≥98.0% (GC,T) | |
4-Chloro-3-(trifluoromethyl)benzoic Acid |
Safety and Handling
TSCA : No