Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-3-(trifluoromethyl)benzoic acid, 97+%

$98.66 - $350.96

Chemical Identifiers

Chemical Name or Material 4-Chloro-3-(trifluoromethyl)benzoic acid
CAS 1737-36-6
Molecular Formula C8H4ClF3O2
Formula Weight 224.57
MDL Number MFCD01631468
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2365603

 
alfa aesar™
B2365603
1g Each for $98.66

AAB2365606

 
alfa aesar™
B2365606
5g Each for $350.96
Description & Specifications

Specifications

Formula Weight 224.57
Chemical Name or Material 4-Chloro-3-(trifluoromethyl)benzoic acid
Assay ≥97%
Molecular Formula C8H4ClF3O2
MDL Number MFCD01631468
CAS 1737-36-6
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 163° to 165°C
Quantity 1g

Applications
It is used in the synthesis of pyrrolo[2,3-d]pyrimidine derivatives and their antiproliferative activity against melanoma cell line.

Solubility
Slightly soluble in water at 25°C 0.039 g/L.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.