missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Chloro-3-(trifluoromethyl)benzenesulfonyl chloride, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC450310010

 View more versions of this product

Catalog No. AC450310010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

4-Chloro-3-(trifluoromethyl)benzenesulfonyl chloride
28.0°C to 30.0°C
32333-53-2
C7H3Cl2F3O2S
SSULGNXFUGLULI-UHFFFAOYSA-N
4-chloro-3-(trifluoromethyl)benzenesulfonyl chloride
2780719
97%
Glass Bottle
1g
96% min. (GC)
4-chloro-3-trifluoromethyl benzene-1-sulfonyl chloride, 4-chloro-3-trifluoromethyl benzenesulfonyl chlori, 4-chloro-3-trifluoromethyl benzenesulfonyl chloride, 4-chloro-3-trifluoromethyl benzenesulfonylchloride, 4-chloro-3-trifluoromethyl benzenesulphonyl chloride, 4-chloro-3-trifluoromethyl-benzenesulfonyl chloride, 4-chloro-3-trifluoromethylbenzenesulfonylchloride, 4-chloro-5-trifluoromethyl benzenesulfonyl chloride, benzenesulfonyl chloride, 4-chloro-3-trifluoromethyl, ksc222e0j
C1=CC(=C(C=C1S(=O)(=O)Cl)C(F)(F)F)Cl
279.06
279.06

Chemical Identifiers

32333-53-2
279.06
4-chloro-3-trifluoromethyl benzene-1-sulfonyl chloride, 4-chloro-3-trifluoromethyl benzenesulfonyl chlori, 4-chloro-3-trifluoromethyl benzenesulfonyl chloride, 4-chloro-3-trifluoromethyl benzenesulfonylchloride, 4-chloro-3-trifluoromethyl benzenesulphonyl chloride, 4-chloro-3-trifluoromethyl-benzenesulfonyl chloride, 4-chloro-3-trifluoromethylbenzenesulfonylchloride, 4-chloro-5-trifluoromethyl benzenesulfonyl chloride, benzenesulfonyl chloride, 4-chloro-3-trifluoromethyl, ksc222e0j
4-chloro-3-(trifluoromethyl)benzenesulfonyl chloride
C7H3Cl2F3O2S
SSULGNXFUGLULI-UHFFFAOYSA-N
2780719
C1=CC(=C(C=C1S(=O)(=O)Cl)C(F)(F)F)Cl
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

SDS Documents