missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Chloro-3-(trifluoromethyl)benzeneboronic acid, 96%, Thermo Scientific Chemicals
$146.49 - $543.81
Chemical Identifiers
CAS | 176976-42-4 |
---|---|
Molecular Formula | C7H5BClF3O2 |
Molecular Weight (g/mol) | 224.37 |
MDL Number | MFCD03094999 |
InChI Key | XHKNQBNGLMOTDB-UHFFFAOYSA-N |
Synonym | 4-chloro-3-trifluoromethyl phenylboronic acid, 4-chloro-3-trifluoromethyl benzeneboronic acid, 4-chloro-3-trifluoromethylphenylboronic acid, 4-chloro-3-trifluoromethyl phenyl boronic acid, 4-chloro-3-trifluoromethyl phenylboronicacid, 4-chloro-3-trifluoromethylphenyl boronic acid, boronic acid, 4-chloro-3-trifluoromethyl phenyl, pubchem1796, acmc-1cg57 |
PubChem CID | 2782671 |
IUPAC Name | [4-chloro-3-(trifluoromethyl)phenyl]boronic acid |
SMILES | B(C1=CC(=C(C=C1)Cl)C(F)(F)F)(O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL2010503
|
Thermo Scientific Chemicals
L2010503 |
1 g |
Each for $146.49
|
|
|||||
AAL2010506
|
Thermo Scientific Chemicals
L2010506 |
5 g |
Each for $543.81
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
176976-42-4 | |
224.37 | |
XHKNQBNGLMOTDB-UHFFFAOYSA-N | |
2782671 | |
B(C1=CC(=C(C=C1)Cl)C(F)(F)F)(O)O |
C7H5BClF3O2 | |
MFCD03094999 | |
4-chloro-3-trifluoromethyl phenylboronic acid, 4-chloro-3-trifluoromethyl benzeneboronic acid, 4-chloro-3-trifluoromethylphenylboronic acid, 4-chloro-3-trifluoromethyl phenyl boronic acid, 4-chloro-3-trifluoromethyl phenylboronicacid, 4-chloro-3-trifluoromethylphenyl boronic acid, boronic acid, 4-chloro-3-trifluoromethyl phenyl, pubchem1796, acmc-1cg57 | |
[4-chloro-3-(trifluoromethyl)phenyl]boronic acid |
Specifications
176976-42-4 | |
MFCD03094999 | |
4-chloro-3-trifluoromethyl phenylboronic acid, 4-chloro-3-trifluoromethyl benzeneboronic acid, 4-chloro-3-trifluoromethylphenylboronic acid, 4-chloro-3-trifluoromethyl phenyl boronic acid, 4-chloro-3-trifluoromethyl phenylboronicacid, 4-chloro-3-trifluoromethylphenyl boronic acid, boronic acid, 4-chloro-3-trifluoromethyl phenyl, pubchem1796, acmc-1cg57 | |
B(C1=CC(=C(C=C1)Cl)C(F)(F)F)(O)O | |
224.37 | |
224.38 | |
4-Chloro-3-(trifluoromethyl)benzeneboronic acid |
C7H5BClF3O2 | |
1 g | |
XHKNQBNGLMOTDB-UHFFFAOYSA-N | |
[4-chloro-3-(trifluoromethyl)phenyl]boronic acid | |
2782671 | |
96% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only