Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-3-(trifluoromethyl)benzaldehyde, 97%

$226.01 - $226.01

Chemical Identifiers

Chemical Name or Material 4-Chloro-3-(trifluoromethyl)benzaldehyde
CAS 34328-46-6
Molecular Formula C8H4ClF3O
Formula Weight 208.57
MDL Number MFCD00040722
Beilstein 2101897
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2402206

  SDS
alfa aesar™
B2402206
5g Each for $226.01
Description & Specifications

Specifications

Formula Weight 208.57
Boiling Point 54°C (10mmHg)
Molecular Formula C8H4ClF3O
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 5g
MDL Number MFCD00040722
Beilstein 2101897
CAS 34328-46-6
Chemical Name or Material 4-Chloro-3-(trifluoromethyl)benzaldehyde
Assay 97%
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
4-Chloro-3-(trifluoromethyl)benzaldehyde is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.