Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-Chloro-3'-(trifluoromethyl)acetophenone, 99%

$172.41 - $694.22

Chemical Identifiers

Chemical Name or Material 4'-Chloro-3'-(trifluoromethyl)acetophenone
CAS 129825-11-2
Molecular Formula C9H6ClF3O
Formula Weight 222.6
MDL Number MFCD03094146
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1988403

 
alfa aesar™
L1988403
1g Each for $172.41

AAL1988406

 
alfa aesar™
L1988406
5g Each for $694.22
Description & Specifications

Specifications

Formula Weight 222.6
Chemical Name or Material 4'-Chloro-3'-(trifluoromethyl)acetophenone
Molecular Formula C9H6ClF3O
CAS 129825-11-2
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 99%
MDL Number MFCD03094146
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 60° to 63°C
Quantity 1g
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.