Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-3-(trifluoromethoxy)phenylacetic acid, 97%

$108.97 - $280.15

Chemical Identifiers

Chemical Name or Material 4-Chloro-3-(trifluoromethoxy)phenylacetic acid
CAS 886501-02-6
Molecular Formula C9H6ClF3O3
Formula Weight 254.59
MDL Number MFCD06660251
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2668203

 
alfa aesar™
H2668203
1g Each for $280.15

AAH26682MD

 
alfa aesar™
H26682MD
250mg Each for $108.97
Description & Specifications

Specifications

Formula Weight 254.59
Chemical Name or Material 4-Chloro-3-(trifluoromethoxy)phenylacetic acid
Assay 97%
Molecular Formula C9H6ClF3O3
MDL Number MFCD06660251
CAS 886501-02-6
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 61° to 63°C
Quantity 1g

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.