Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-3-(trifluoromethoxy)phenol, 97%

$226.01 - $226.01

Chemical Identifiers

Chemical Name or Material 4-Chloro-3-(trifluoromethoxy)phenol
CAS 886500-85-2
Molecular Formula C7H4ClF3O2
Formula Weight 212.56
MDL Number MFCD06660247
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2677203

  SDS
alfa aesar™
H2677203
1g Each for $226.01
Description & Specifications

Specifications

Formula Weight 212.56
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 4-Chloro-3-(trifluoromethoxy)phenol
Quantity 1g
Health Hazard 1 Danger
Assay 97%
CAS 886500-85-2
MDL Number MFCD06660247
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Molecular Formula C7H4ClF3O2
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.