missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-3-(trifluoromethoxy)benzoic acid, 97%

$161.62 - $480.26

Chemical Identifiers

CAS 886500-50-1
Molecular Formula C8H4ClF3O3
Formula Weight 240.57
MDL Number MFCD06660238
Chemical Name or Material 4-Chloro-3-(trifluoromethoxy)benzoic acid
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2685403 SDS Alfa Aesar™
H2685403
1g Each for $161.62
AAH2685406 SDS Alfa Aesar™
H2685406
5g Each for $480.26
Specifications

Description

Applications
4-Chloro-3-(trifluoromethoxy)benzoic acid is used as pharmaceuticals intermediates.

Solubility
Sparingly Soluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Description & Specifications

Chemical Identifiers

886500-50-1
240.57
4-Chloro-3-(trifluoromethoxy)benzoic acid
C8H4ClF3O3
MFCD06660238

Specifications

123° to 126°C
886500-50-1
240.57
4-Chloro-3-(trifluoromethoxy)benzoic acid
1g
C8H4ClF3O3
MFCD06660238
97%

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning