Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-3-(trifluoromethoxy)aniline, 97%

Chemical Identifiers

Chemical Name or Material 4-Chloro-3-(trifluoromethoxy)aniline
CAS 97608-50-9
Molecular Formula C7H5ClF3NO
Formula Weight 211.57
MDL Number MFCD09832255
UN Number UN2810
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3175203

 
alfa aesar™
H3175203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH31752MD

 
alfa aesar™
H31752MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 211.57
Chemical Name or Material 4-Chloro-3-(trifluoromethoxy)aniline
Assay 97%
Molecular Formula C7H5ClF3NO
MDL Number MFCD09832255
CAS 97608-50-9
UN Number UN2810
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
Quantity 1g

Applications
4-Chloro-3-(trifluoromethoxy)aniline is widely utilized as an intermediate in organic synthesis.

Notes
Store in a cool and dry place. Keep the container tightly sealed.

Safety and Handling

Danger

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.