missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-3-pyridinesulfonamide, 98%

$39.88 - $107.30

Chemical Identifiers

CAS 33263-43-3
Molecular Formula C5H5ClN2O2S
Formula Weight 192.62
MDL Number MFCD04038136
Chemical Name or Material 4-Chloro-3-pyridinesulfonamide
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1949306 SDS Alfa Aesar™
L1949306
5g Each for $107.30
AAL1949303 SDS Alfa Aesar™
L1949303
1g Each for $39.88
Specifications Description & Specifications

Chemical Identifiers

33263-43-3
192.62
4-Chloro-3-pyridinesulfonamide
C5H5ClN2O2S
MFCD04038136

Specifications

150° to 155°C
33263-43-3
192.62
4-Chloro-3-pyridinesulfonamide
5g
C5H5ClN2O2S
MFCD04038136
98%

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

Recommended Storage : 2° to 8°C