Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-3-nitropyridine hydrochloride, 90+%

$62.53 - $188.35

Chemical Identifiers

Chemical Name or Material 4-Chloro-3-nitropyridine hydrochloride
CAS 54079-68-4
Molecular Formula C5H3ClN2O2·HCl
Formula Weight 195
MDL Number MFCD02683026
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1972803

  SDS
alfa aesar™
L1972803
1g Each for $62.53

AAL1972806

  SDS
alfa aesar™
L1972806
5g Each for $188.35
Description & Specifications

Specifications

Formula Weight 195
Chemical Name or Material 4-Chloro-3-nitropyridine hydrochloride
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
MDL Number MFCD02683026
CAS 54079-68-4
Molecular Formula C5H3ClN2O2·HCl
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Melting Point 135° to 138°C
Quantity 1g
Assay ≥90%
Safety and Handling

Warning

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.