missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-3-nitropyridine, 95%, may cont. variable amounts of oligomers

$138.08 - $494.11

Chemical Identifiers

CAS 13091-23-1
Molecular Formula C5H3ClN2O2
Formula Weight 158.54
MDL Number MFCD02683025
Chemical Name or Material 4-Chloro-3-nitropyridine
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1936414 SDS Alfa Aesar™
L1936414
25g Each for $494.11
AAL1936406 SDS Alfa Aesar™
L1936406
5g Each for $138.08
Specifications

Description

Applications
4-Chloro-3-nitropyridine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Description & Specifications

Chemical Identifiers

13091-23-1
158.54
4-Chloro-3-nitropyridine
C5H3ClN2O2
MFCD02683025

Specifications

≈32°C
13091-23-1
158.54
4-Chloro-3-nitropyridine
25g
C5H3ClN2O2
MFCD02683025
95%, may cont. variable amounts of oligomers

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

Recommended Storage : −30° to −10°C