Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-3-nitrophenyl isocyanate, 94%

Chemical Identifiers

Chemical Name or Material 4-Chloro-3-nitrophenyl isocyanate
CAS 40397-96-4
Molecular Formula C7H3ClN2O3
Formula Weight 198.57
MDL Number MFCD00037062
UN Number UN2811
Beilstein 2807129
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1108703

  SDS
alfa aesar™
L1108703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1108706

  SDS
alfa aesar™
L1108706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 198.57
Chemical Name or Material 4-Chloro-3-nitrophenyl isocyanate
CAS 40397-96-4
Health Hazard 1 Danger
MDL Number MFCD00037062
Assay 94%
Health Hazard 2 GHS H Statement
H331-H334-H302-H312-H315-H319-H317-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Beilstein 2807129
Quantity 1g
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Melting Point 63° to 69°C
Recommended Storage 2° to 8°C
Health Hazard 3 GHS P Statement
P284a-P261-P280-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C7H3ClN2O3
UN Number UN2811
Safety and Handling

Danger

GHS P Statement
P284a-P261-P280-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H334-H302-H312-H315-H319-H317-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.

2° to 8°C