missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-3-nitrophenyl isocyanate, 94%

Manufacturer:  Alfa Aesar™ L1108706

 View more versions of this product

Catalog No. AAL1108706



Specifications

Specifications

4-Chloro-3-nitrophenyl isocyanate
5g
C7H3ClN2O3
UN2811
4-chloro-3-nitrophenyl isocyanate, 4-chloro-3-nitrophenylisocyanate, 3-nitro-4-chlorophenyl isocyanate, 4-chloro-3-nitrobenzenisocyanate, acmc-209jdv, #, benzene,1-chloro-4-isocyanato-2-nitro, 1-chloranyl-4-isocyanato-2-nitro-benzene, isocyanic acid 4-chloro-3-nitrophenyl ester
C1=CC(=C(C=C1N=C=O)[N+](=O)[O-])Cl
198.562
198.57
63°C to 69°C
40397-96-4
MFCD00037062
2807129
ZCPHLSPLEGBTCZ-UHFFFAOYSA-N
1-chloro-4-isocyanato-2-nitrobenzene
142448
94%

Chemical Identifiers

40397-96-4
198.562
ZCPHLSPLEGBTCZ-UHFFFAOYSA-N
142448
C1=CC(=C(C=C1N=C=O)[N+](=O)[O-])Cl
C7H3ClN2O3
MFCD00037062
4-chloro-3-nitrophenyl isocyanate, 4-chloro-3-nitrophenylisocyanate, 3-nitro-4-chlorophenyl isocyanate, 4-chloro-3-nitrobenzenisocyanate, acmc-209jdv, #, benzene,1-chloro-4-isocyanato-2-nitro, 1-chloranyl-4-isocyanato-2-nitro-benzene, isocyanic acid 4-chloro-3-nitrophenyl ester
1-chloro-4-isocyanato-2-nitrobenzene
Safety and Handling

Safety and Handling

GHS H Statement
H331-H334-H302-H312-H315-H319-H317-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.

GHS P Statement
P284a-P261-P280-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

Recommended Storage : 2° to 8°C

Documents