missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-chloro-3-nitrophenol, 98%, ACROS Organics™

Chemical Identifiers

CAS 610-78-6
Molecular Formula C6H4ClNO3
Molecular Weight (g/mol) 173.56
MDL Number MFCD00043546
InChI Key JUIKCULGDIZNDI-UHFFFAOYSA-N
Synonym 2-chloro-5-hydroxy nitrobenzene
PubChem CID 69127
IUPAC Name 4-chloro-3-nitrophenol
SMILES C1=CC(=C(C=C1O)[N+](=O)[O-])Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC369590250 SDS View Documents Thermo Fisher Scientific
AC369590250
25g Glass bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

610-78-6
173.56
JUIKCULGDIZNDI-UHFFFAOYSA-N
69127
C1=CC(=C(C=C1O)[N+](=O)[O-])Cl
C6H4ClNO3
MFCD00043546
2-chloro-5-hydroxy nitrobenzene
4-chloro-3-nitrophenol

Specifications

Authentic
Yellow
25g
97.5
97.5% min. (GC)
ClC6H3(NO2)OH
06, IV, 1352
JUIKCULGDIZNDI-UHFFFAOYSA-N
4-chloro-3-nitrophenol
69127
Crystalline Powder
4-chloro-3-nitrophenol
Glass bottle
125°C to 129°C
610-78-6
100.0
C6H4ClNO3
MFCD00043546
2-chloro-5-hydroxy nitrobenzene
C1=CC(=C(C=C1O)[N+](=O)[O-])Cl
173.56
173.56
98%
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning