Learn More
Alfa Aesar™ 4-Chloro-3-nitrocinnamic acid, 98%
Manufacturer: Alfa Aesar™ A1495414
Specifications
4-Chloro-3-nitrocinnamic acid | |
25g | |
C9H6ClNO4 | |
4-chloro-3-nitrocinnamic acid, trans-4-chloro-3-nitrocinnamic acid, 3-4-chloro-3-nitrophenyl acrylic acid, e-3-4-chloro-3-nitrophenyl acrylic acid, 2e-3-4-chloro-3-nitrophenyl prop-2-enoic acid, e-3-4-chloro-3-nitrophenyl prop-2-enoic acid, 4-chloro-3-nitro-cinnamic acid, 3-nitro-4-chlorocinnamic acid, qbdaltimhoitiu-duxpyhpusa, 3-4-chloro-3-nitrophenyl propenoic acid | |
C1=CC(=C(C=C1C=CC(=O)O)[N+](=O)[O-])Cl | |
688108 | |
98% |
186°C to 188°C | |
20797-48-2 | |
MFCD00063311 | |
QBDALTIMHOITIU-DUXPYHPUSA-N | |
227.6 | |
227.61 |
Chemical Identifiers
20797-48-2 | |
227.6 | |
QBDALTIMHOITIU-DUXPYHPUSA-N | |
688108 |
C9H6ClNO4 | |
MFCD00063311 | |
4-chloro-3-nitrocinnamic acid, trans-4-chloro-3-nitrocinnamic acid, 3-4-chloro-3-nitrophenyl acrylic acid, e-3-4-chloro-3-nitrophenyl acrylic acid, 2e-3-4-chloro-3-nitrophenyl prop-2-enoic acid, e-3-4-chloro-3-nitrophenyl prop-2-enoic acid, 4-chloro-3-nitro-cinnamic acid, 3-nitro-4-chlorocinnamic acid, qbdaltimhoitiu-duxpyhpusa, 3-4-chloro-3-nitrophenyl propenoic acid | |
C1=CC(=C(C=C1C=CC(=O)O)[N+](=O)[O-])Cl |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning