missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Chloro-3-nitrobenzotrifluoride 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 109700100

 View more versions of this product

Catalog No. AC109700100

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

4-Chloro-3-nitrobenzotrifluoride
121-17-5
Authentic
Glass bottle
10g
Liquid
98%
ClC6H3(NO2)CF3
MFCD00007084
98%
97.5
100.0
1.4883 to 1.4903 (20°C, 589nm)
Clear yellow
≥97.5%
C7H3ClF3NO2
225.55
4-Chloro-3-nitro-α, α, α-trifluorotoluene

Chemical Identifiers

121-17-5
100.0
C7H3ClF3NO2
225.55
4-Chloro-3-nitro-α, α, α-trifluorotoluene
≥97.5%
97.5
98%
ClC6H3(NO2)CF3
MFCD00007084
4-Chloro-3-nitrobenzotrifluoride, 98%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 204-451-6

RTECSNumber : XS9140000

TSCA : TSCA