Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-3-nitrobenzotrifluoride, 98%

$46.80 - $46.80

Chemical Identifiers

Chemical Name or Material 4-Chloro-3-nitrobenzotrifluoride
CAS 121-17-5
Molecular Formula C7H3ClF3NO2
Formula Weight 225.55
MDL Number MFCD00007084
UN Number UN2307
Beilstein 523986
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1148822

  SDS
alfa aesar™
A1148822
100g Each for $46.80

AAA114880E

  SDS
alfa aesar™
A114880E
2500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1148836

  SDS
alfa aesar™
A1148836
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 225.55
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 4-Chloro-3-nitrobenzotrifluoride
TSCA TSCA
Assay 98%
Molecular Formula C7H3ClF3NO2
MDL Number MFCD00007084
CAS 121-17-5
UN Number UN2307
Beilstein 523986
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: 3-NITRO-4-CHLOROBENZOTRIFLUORIDE
RTECS Number XS9140000
Flash Point 101°C (213°F)
Boiling Point 221° to 223°C
Melting Point −3°C
Quantity 100g
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.