Please login to your online account to display your discounted pricing

4-Chloro-3-nitrobenzoic Acid 99.5%, ACROS Organics™


Quantity 500g

Manufacturer: thermo fisher scientific  109675000

 View more versions of this product

Catalog No. AC109675000

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Powder
Formula Weight 201.57
Assay ≥99.4%
Quantity 500g
Color Light yellow to cream
Melting Point 181° to 185°C
TSCA TSCA
RTECS Number DG5425050
EINECS Number 202-550-9
Beilstein 09, 402
CAS Max % 100.0
Water 0.5% max. (K.F.)
Molecular Formula C7H4ClNO4
Name Note 99.5%
CAS 96-99-1
Linear Formula ClC6H3(NO2)CO2H
MDL Number MFCD00007079
CAS Min % 99.4
Chemical Name or Material 4-Chloro-3-nitrobenzoic acid
Assay Percent Range 99.5%
Packaging Plastic bottle
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 4-Chloro-3-nitrobenzoic acid
Name Note 99.5%
CAS 96-99-1
CAS Min % 99.4
CAS Max % 100.0
Assay ≥99.4%
Assay Percent Range 99.5%
Molecular Formula C7H4ClNO4
Linear Formula ClC6H3(NO2)CO2H
Formula Weight 201.57
MDL Number MFCD00007079
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents