missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Chloro-3-nitrobenzoic Acid 99.5%, ACROS Organics™

$112.26 - $112.26

Chemical Identifiers

CAS 96-99-1
Molecular Formula C7H4ClNO4
Molecular Weight (g/mol) 201.562
MDL Number MFCD00007079
InChI Key DFXQXFGFOLXAPO-UHFFFAOYSA-N
Synonym 4-chloro-3-nitrobenzoic acid, 3-nitro-4-chlorobenzoic acid, benzoic acid, 4-chloro-3-nitro, kyselina 4-chloro-3-nitrobenzoova, unii-0mlz915q9k, ccris 3126, kyselina 4-chloro-3-nitrobenzoova czech, 4-chloro-3-nitrobenzoic acic, 4-chloro-3-nitro-benzoic acid, 5-carboxy-2-chloronitrobenzene
PubChem CID 7320
SMILES C1=CC(=C(C=C1C(=O)O)[N+](=O)[O-])Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC109675000 SDS View Documents Acros Organics
109675000
500g Each for $112.26
Specifications

Chemical Identifiers

96-99-1
201.562
DFXQXFGFOLXAPO-UHFFFAOYSA-N
7320
C7H4ClNO4
MFCD00007079
4-chloro-3-nitrobenzoic acid, 3-nitro-4-chlorobenzoic acid, benzoic acid, 4-chloro-3-nitro, kyselina 4-chloro-3-nitrobenzoova, unii-0mlz915q9k, ccris 3126, kyselina 4-chloro-3-nitrobenzoova czech, 4-chloro-3-nitrobenzoic acic, 4-chloro-3-nitro-benzoic acid, 5-carboxy-2-chloronitrobenzene
C1=CC(=C(C=C1C(=O)O)[N+](=O)[O-])Cl

Specifications

Authentic
0.5% max. (K.F.)
181°C to 185°C
96-99-1
100.0
C7H4ClNO4
MFCD00007079
4-chloro-3-nitrobenzoic acid, 3-nitro-4-chlorobenzoic acid, benzoic acid, 4-chloro-3-nitro, kyselina 4-chloro-3-nitrobenzoova, unii-0mlz915q9k, ccris 3126, kyselina 4-chloro-3-nitrobenzoova czech, 4-chloro-3-nitrobenzoic acic, 4-chloro-3-nitro-benzoic acid, 5-carboxy-2-chloronitrobenzene
C1=CC(=C(C=C1C(=O)O)[N+](=O)[O-])Cl
7320
Powder
4-Chloro-3-nitrobenzoic acid, 99.5%
Plastic bottle
Cream to Yellow
500g
99.4
99.5%
ClC6H3(NO2)CO2H
09, 402
DFXQXFGFOLXAPO-UHFFFAOYSA-N
201.562
201.57
≥99.4%
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 202-550-9

RTECSNumber : DG5425050

TSCA : TSCA