Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-3-nitrobenzeneboronic acid, 97%

$111.65 - $233.55

Chemical Identifiers

Chemical Name or Material 4-Chloro-3-nitrobenzeneboronic acid
CAS 151169-67-4
Molecular Formula C6H5BClNO4
Formula Weight 201.37
MDL Number MFCD02258950
Synonym 4-Chloro-3-nitrophenylboronic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH27093MC

  SDS
alfa aesar™
H27093MC
100mg Each for $111.65

AAH27093ME

  SDS
alfa aesar™
H27093ME
500mg Each for $233.55
Description & Specifications

Specifications

Formula Weight 201.37
Health Hazard 1 Warning
CAS 151169-67-4
Melting Point ≈275°C
Quantity 100mg
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 4-Chloro-3-nitrobenzeneboronic acid
Synonym 4-Chloro-3-nitrophenylboronic acid
Assay 97%
Molecular Formula C6H5BClNO4
MDL Number MFCD02258950
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.