Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-3-nitrobenzamide, 98%

$44.00 - $162.73

Chemical Identifiers

Chemical Name or Material 4-Chloro-3-nitrobenzamide
CAS 16588-06-0
Molecular Formula C7H5ClN2O3
Formula Weight 200.58
MDL Number MFCD00017134
Beilstein 645210
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1403403

  SDS
alfa aesar™
L1403403
1g Each for $44.00

AAL1403406

  SDS
alfa aesar™
L1403406
5g Each for $162.73
Description & Specifications

Specifications

Formula Weight 200.58
TSCA TSCA
MDL Number MFCD00017134
CAS 16588-06-0
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 150° to 154°C
Chemical Name or Material 4-Chloro-3-nitrobenzamide
Assay 98%
Molecular Formula C7H5ClN2O3
Beilstein 645210
RTECS Number CV2465000
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.