Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-3-nitroanisole, 98%

$170.27 - $715.72

Chemical Identifiers

Chemical Name or Material 4-Chloro-3-nitroanisole
CAS 10298-80-3
Molecular Formula C7H6ClNO3
Formula Weight 187.58
MDL Number MFCD00007077
Beilstein 640872
Synonym 1-Chloro-4-methoxy-2-nitrobenzene; 4-Methoxy-2-nitrochlorobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1228222

  SDS
alfa aesar™
A1228222
100g Each for $170.27

AAA1228236

  SDS
alfa aesar™
A1228236
500g Each for $715.72
Description & Specifications

Specifications

Formula Weight 187.58
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD00007077
Melting Point 42° to 45°C
Assay 98%
Beilstein 640872
RTECS Number BZ8580000
Quantity 100g
Chemical Name or Material 4-Chloro-3-nitroanisole
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 10298-80-3
Flash Point >110°C (230°F)
Molecular Formula C7H6ClNO3
Synonym 1-Chloro-4-methoxy-2-nitrobenzene; 4-Methoxy-2-nitrochlorobenzene
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.