Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-Chloro-3'-nitroacetophenone, 97%

$65.09 - $721.74

Chemical Identifiers

Chemical Name or Material 4'-Chloro-3'-nitroacetophenone
CAS 5465-65-6
Molecular Formula C8H6ClNO3
Formula Weight 199.6
MDL Number MFCD00007083
Beilstein 1640627
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1512106

  SDS
alfa aesar™
A1512106
5g Each for $65.09

AAA1512114

  SDS
alfa aesar™
A1512114
25g Each for $232.04

AAA1512122

  SDS
alfa aesar™
A1512122
100g Each for $721.74
Description & Specifications

Specifications

Formula Weight 199.6
Chemical Name or Material 4'-Chloro-3'-nitroacetophenone
Assay 97%
Molecular Formula C8H6ClNO3
MDL Number MFCD00007083
CAS 5465-65-6
Beilstein 1640627
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 96° to 100°C
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.