missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-Chloro-3'-nitroacetophenone, 97%

Manufacturer:  Alfa Aesar™ A1512114

 View more versions of this product

Catalog No. AAA1512114



Specifications

Specifications

4'-Chloro-3'-nitroacetophenone
25g
C8H6ClNO3
1640627
YEVPHFIFGUWSMG-UHFFFAOYSA-N
1-(4-chloro-3-nitrophenyl)ethanone
79596
97%
96°C to 100°C
5465-65-6
MFCD00007083
4'-chloro-3'-nitroacetophenone, 1-4-chloro-3-nitrophenyl ethanone, 4-chloro-3-nitroacetophenone, ethanone, 1-4-chloro-3-nitrophenyl, 1-4-chloro-3-nitrophenyl ethan-1-one, unii-gkv85f2zxl, acetophenone, 4'-chloro-3'-nitro, gkv85f2zxl, 4-chloro-3'-nitroacetophenone, 3'-nitro-4'-chloro acetophenone
CC(=O)C1=CC(=C(C=C1)Cl)[N+](=O)[O-]
199.59
199.6

Chemical Identifiers

5465-65-6
199.59
YEVPHFIFGUWSMG-UHFFFAOYSA-N
79596
CC(=O)C1=CC(=C(C=C1)Cl)[N+](=O)[O-]
C8H6ClNO3
MFCD00007083
4'-chloro-3'-nitroacetophenone, 1-4-chloro-3-nitrophenyl ethanone, 4-chloro-3-nitroacetophenone, ethanone, 1-4-chloro-3-nitrophenyl, 1-4-chloro-3-nitrophenyl ethan-1-one, unii-gkv85f2zxl, acetophenone, 4'-chloro-3'-nitro, gkv85f2zxl, 4-chloro-3'-nitroacetophenone, 3'-nitro-4'-chloro acetophenone
1-(4-chloro-3-nitrophenyl)ethanone
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents