missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-methylpyridine hydrochloride, 97%, Thermo Scientific Chemicals

$197.71 - $197.71

Chemical Identifiers

CAS 19524-08-4
Molecular Formula C6H7Cl2N
Molecular Weight (g/mol) 164.029
MDL Number MFCD03092890
InChI Key WPIUSHWGBZMBHZ-UHFFFAOYSA-N
Synonym 4-chloro-3-methylpyridine hydrochloride, 4-chloro-3-picoline hcl, 4-chloro-3-methylpyridine hcl, 4-chloro-3-picoline hydrochloride, pyridine, 4-chloro-3-methyl-, hydrochloride, 4-chloro-3-methylpyridinehydrochloride, acmc-20amlc, pubchem9218, ksc495e6r, 4-chloro-3methylpyridine hydrochloride
PubChem CID 16217655
IUPAC Name 4-chloro-3-methylpyridine;hydrochloride
SMILES CC1=C(C=CN=C1)Cl.Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1955803
View Documents
Thermo Scientific Chemicals
L1955803
1 g N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

19524-08-4
164.029
WPIUSHWGBZMBHZ-UHFFFAOYSA-N
16217655
CC1=C(C=CN=C1)Cl.Cl
C6H7Cl2N
MFCD03092890
4-chloro-3-methylpyridine hydrochloride, 4-chloro-3-picoline hcl, 4-chloro-3-methylpyridine hcl, 4-chloro-3-picoline hydrochloride, pyridine, 4-chloro-3-methyl-, hydrochloride, 4-chloro-3-methylpyridinehydrochloride, acmc-20amlc, pubchem9218, ksc495e6r, 4-chloro-3methylpyridine hydrochloride
4-chloro-3-methylpyridine;hydrochloride

Specifications

19524-08-4
Odorless
MFCD03092890
107953
WPIUSHWGBZMBHZ-UHFFFAOYSA-N
4-chloro-3-methylpyridine;hydrochloride
16217655
97%
∼185°C
C6H7Cl2N
1 g
4-chloro-3-methylpyridine hydrochloride, 4-chloro-3-picoline hcl, 4-chloro-3-methylpyridine hcl, 4-chloro-3-picoline hydrochloride, pyridine, 4-chloro-3-methyl-, hydrochloride, 4-chloro-3-methylpyridinehydrochloride, acmc-20amlc, pubchem9218, ksc495e6r, 4-chloro-3methylpyridine hydrochloride
CC1=C(C=CN=C1)Cl.Cl
164.029
164.04
4-Chloro-3-methylpyridine hydrochloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only