Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-3-methylphenol, 99%

Chemical Identifiers

Chemical Name or Material 4-Chloro-3-methylphenol
CAS 59-50-7
Molecular Formula C7H7ClO
Formula Weight 142.59
MDL Number MFCD00002323
UN Number UN3437
Beilstein 1237629
Merck Index 14,2133
Synonym 4-Chloro-m-cresol; PCMC
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1782336

 
alfa aesar™
A1782336
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1782322

 
alfa aesar™
A1782322
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 142.59
Chemical Name or Material 4-Chloro-3-methylphenol
TSCA TSCA
Assay 99%
Molecular Formula C7H7ClO
MDL Number MFCD00002323
CAS 59-50-7
UN Number UN3437
Beilstein 1237629
Merck Index 14,2133
Synonym 4-Chloro-m-cresol; PCMC
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H318-H302-H312-H317
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
May cause an allergic skin reaction.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P310-P301+P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: CHLOROCRESOLS, SOLID
RTECS Number GO7100000
Flash Point 118°C (244°F)
Boiling Point 234° to 236°C
Melting Point 64° to 68°C
Quantity 500g
Safety and Handling

Danger

GHS P Statement
P261-P280-P305+P351+P338-P310-P301+P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H318-H302-H312-H317
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
May cause an allergic skin reaction.