Please login to your online account to display your discounted pricing

4-chloro-3-methylbenzylamine hydrochloride, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Chloro-3-methylbenzylamine hydrochloride
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C8H9N·ClH
Formula Weight 192.09
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC429660010

  SDS
thermo fisher scientific
AC429660010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC429660050

  SDS
thermo fisher scientific
AC429660050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder
Assay ≥96.0%
Formula Weight 192.09
Quantity 1g
Color White to yellow to pink
Chemical Name or Material 4-Chloro-3-methylbenzylamine hydrochloride
Molecular Formula C8H9N·ClH
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 1264198-68-6
Assay Percent Range 97%
Infrared Spectrum Authentic
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.