Learn More
4-Chloro-3-methylbenzoyl chloride, 96%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H6610903
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
4-Chloro-3-methylbenzoyl chloride | |
Colorless to Yellow | |
MFCD11111062 | |
UN3265 | |
3-methyl-4-chloro benzoylchloride, 4-chloro-3-methylbenzoylchloride, 3-methyl-4-chlorobenzoyl chloride, 4-chloro-3-methyl-benzoyl chloride, 4-chloranyl-3-methyl-benzoyl chloride, benzoyl chloride, 4-chloro-3-methyl | |
CC1=C(C=CC(=C1)C(=O)Cl)Cl | |
189.035 | |
189.03 | |
Liquid |
21900-24-3 | |
C8H6Cl2O | |
1 g | |
Moisture sensitive | |
NECBSSKNQDXVKN-UHFFFAOYSA-N | |
4-chloro-3-methylbenzoyl chloride | |
13058375 | |
96% |
Chemical Identifiers
21900-24-3 | |
189.035 | |
NECBSSKNQDXVKN-UHFFFAOYSA-N | |
13058375 | |
CC1=C(C=CC(=C1)C(=O)Cl)Cl |
C8H6Cl2O | |
MFCD11111062 | |
3-methyl-4-chloro benzoylchloride, 4-chloro-3-methylbenzoylchloride, 3-methyl-4-chlorobenzoyl chloride, 4-chloro-3-methyl-benzoyl chloride, 4-chloranyl-3-methyl-benzoyl chloride, benzoyl chloride, 4-chloro-3-methyl | |
4-chloro-3-methylbenzoyl chloride |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packaging Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen