missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-3-methylbenzoyl chloride, 96%

$49.66 - $157.30

Chemical Identifiers

CAS 21900-24-3
Molecular Formula C8H6Cl2O
Molecular Weight (g/mol) 189.035
MDL Number MFCD11111062
InChI Key NECBSSKNQDXVKN-UHFFFAOYSA-N
Synonym 3-methyl-4-chloro benzoylchloride, 4-chloro-3-methylbenzoylchloride, 3-methyl-4-chlorobenzoyl chloride, 4-chloro-3-methyl-benzoyl chloride, 4-chloranyl-3-methyl-benzoyl chloride, benzoyl chloride, 4-chloro-3-methyl
PubChem CID 13058375
IUPAC Name 4-chloro-3-methylbenzoyl chloride
SMILES CC1=C(C=CC(=C1)C(=O)Cl)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6610903 View Documents Alfa Aesar™
H6610903
1g Each for $157.30
 
AAH66109MD View Documents Alfa Aesar™
H66109MD
250mg Each for $49.66
 
Specifications

Chemical Identifiers

21900-24-3
189.035
NECBSSKNQDXVKN-UHFFFAOYSA-N
13058375
CC1=C(C=CC(=C1)C(=O)Cl)Cl
C8H6Cl2O
MFCD11111062
3-methyl-4-chloro benzoylchloride, 4-chloro-3-methylbenzoylchloride, 3-methyl-4-chlorobenzoyl chloride, 4-chloro-3-methyl-benzoyl chloride, 4-chloranyl-3-methyl-benzoyl chloride, benzoyl chloride, 4-chloro-3-methyl
4-chloro-3-methylbenzoyl chloride

Specifications

Moisture Sensitive
1g
C8H6Cl2O
UN3265
NECBSSKNQDXVKN-UHFFFAOYSA-N
4-chloro-3-methylbenzoyl chloride
13058375
Liquid
4-Chloro-3-methylbenzoyl chloride
Colorless to Yellow
21900-24-3
MFCD11111062
3-methyl-4-chloro benzoylchloride, 4-chloro-3-methylbenzoylchloride, 3-methyl-4-chlorobenzoyl chloride, 4-chloro-3-methyl-benzoyl chloride, 4-chloranyl-3-methyl-benzoyl chloride, benzoyl chloride, 4-chloro-3-methyl
CC1=C(C=CC(=C1)C(=O)Cl)Cl
189.035
189.03
96%
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packaging Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.