Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-3-methylaniline, 98%

$77.73 - $77.73

Chemical Identifiers

Chemical Name or Material 4-Chloro-3-methylaniline
CAS 7149-75-9
Molecular Formula C7H8ClN
Formula Weight 141.6
MDL Number MFCD00066332
UN Number UN2239
Beilstein 636522
Synonym 5-Amino-2-chlorotoluene; 4-Chloro-m-toluidine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2223406

 
alfa aesar™
B2223406
5g Each for $77.73

AAB2223414

 
alfa aesar™
B2223414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 141.6
Chemical Name or Material 4-Chloro-3-methylaniline
Assay 98%
Molecular Formula C7H8ClN
MDL Number MFCD00066332
CAS 7149-75-9
UN Number UN2239
Beilstein 636522
Synonym 5-Amino-2-chlorotoluene; 4-Chloro-m-toluidine
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H331-H373-H302-H312-H315-H319
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P260-P261-P280-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CHLOROTOLUIDINES, SOLID
Melting Point 82° to 85°C
Quantity 5g
Safety and Handling

Danger

GHS P Statement
P260-P261-P280-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H373-H302-H312-H315-H319
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.