missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Chloro-3-methoxybenzoic Acid, TCI America™

Manufacturer:  TCI America C33041G

 View more versions of this product

Catalog No. C33041G



Specifications

Specifications

4-Chloro-3-methoxybenzoic Acid
219°C
85740-98-3
MFCD00269640
OXUUNDMDOOXPKY-UHFFFAOYSA-N
4-chloro-3-methoxybenzoic acid
2063378
Crystal-Powder at 20°C
White
1g
C8H7ClO3
4-Chloro-m-anisic Acid
COC1=C(C=CC(=C1)C(=O)O)Cl
186.591
186.59
≥98.0% (GC,T)

Chemical Identifiers

85740-98-3
186.591
OXUUNDMDOOXPKY-UHFFFAOYSA-N
2063378
COC1=C(C=CC(=C1)C(=O)O)Cl
C8H7ClO3
MFCD00269640
4-Chloro-m-anisic Acid
4-chloro-3-methoxybenzoic acid
Safety and Handling

Safety and Handling

TSCA : No

Documents