Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-3-iodophenol, 98%

$54.03 - $278.61

Chemical Identifiers

Chemical Name or Material 4-Chloro-3-iodophenol
CAS 202982-72-7
Molecular Formula C6H4ClIO
Formula Weight 254.45
MDL Number MFCD00070769
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2554703

  SDS
alfa aesar™
B2554703
1g Each for $54.03

AAB2554709

  SDS
alfa aesar™
B2554709
10g Each for $278.61
Description & Specifications

Specifications

Formula Weight 254.45
Chemical Name or Material 4-Chloro-3-iodophenol
Assay 98%
Molecular Formula C6H4ClIO
MDL Number MFCD00070769
CAS 202982-72-7
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 75° to 77°C
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.