Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-3-iodobenzotrifluoride, 98%

$50.78 - $156.70

Chemical Identifiers

Chemical Name or Material 4-Chloro-3-iodobenzotrifluoride
CAS 672-57-1
Molecular Formula C7H3ClF3I
Formula Weight 306.45
MDL Number MFCD00001037
Beilstein 1870212
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2254814

  SDS
alfa aesar™
B2254814
25g Each for $50.78

AAB2254822

  SDS
alfa aesar™
B2254822
100g Each for $156.70
Description & Specifications

Specifications

Formula Weight 306.45
Flash Point >110°C (230°F)
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Assay 98%
CAS 672-57-1
Beilstein 1870212
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Molecular Formula C7H3ClF3I
Boiling Point 58° to 60°C (0.5mmHg)
MDL Number MFCD00001037
Chemical Name or Material 4-Chloro-3-iodobenzotrifluoride
Quantity 25g
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.