Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-3-(hydroxymethyl)pyridine hydrochloride, 98%

Chemical Identifiers

Chemical Name or Material 4-Chloro-3-(hydroxymethyl)pyridine hydrochloride
CAS 245124-17-8
Formula Weight 180.03
MDL Number MFCD23703142
Beilstein 8339779
Synonym 4-Chloropyridine-3-methanol hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6652203

  SDS
alfa aesar™
H6652203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH66522MD

  SDS
alfa aesar™
H66522MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 180.03
Chemical Name or Material 4-Chloro-3-(hydroxymethyl)pyridine hydrochloride
CAS 245124-17-8
MDL Number MFCD23703142
Assay 98%
Physical Form Solid
Synonym 4-Chloropyridine-3-methanol hydrochloride
Beilstein 8339779
Quantity 1g