Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-3-fluoropyridine hydrochloride, 97%

$52.34 - $220.12

Chemical Identifiers

Chemical Name or Material 4-Chloro-3-fluoropyridine hydrochloride
CAS 119229-74-2
Molecular Formula C5H3ClFN·HCl
Formula Weight 168
MDL Number MFCD03453233
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2718403

 
alfa aesar™
H2718403
1g Each for $52.34

AAH2718406

 
alfa aesar™
H2718406
5g Each for $220.12
Description & Specifications

Specifications

Formula Weight 168
Chemical Name or Material 4-Chloro-3-fluoropyridine hydrochloride
Assay 97%
Molecular Formula C5H3ClFN·HCl
MDL Number MFCD03453233
CAS 119229-74-2
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Melting Point ≈160°C (subl.)
Quantity 1g

Applications
4-Chloro-3-fluoropyridine hydrochloride is employed for the deprotection of phenol ethers.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.