Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-3-fluorophenylacetic acid, 97%

$178.55 - $531.05

Chemical Identifiers

Chemical Name or Material 4-Chloro-3-fluorophenylacetic acid
CAS 883500-51-4
Molecular Formula C8H6ClFO2
Formula Weight 188.59
MDL Number MFCD04115861
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2659203

 
alfa aesar™
H2659203
1g Each for $178.55

AAH2659206

 
alfa aesar™
H2659206
5g Each for $531.05
Description & Specifications

Specifications

Formula Weight 188.59
Chemical Name or Material 4-Chloro-3-fluorophenylacetic acid
Assay 97%
Molecular Formula C8H6ClFO2
MDL Number MFCD04115861
CAS 883500-51-4
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 88° to 91°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.