missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Chloro-3-fluorophenol, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 249970050

 View more versions of this product

Catalog No. AC249970050



Description

Description

Specifications

Specifications

4-Chloro-3-fluorophenol
Glass Bottle
50°C to 55°C
98%
97.5
98%
OHC6H3ClF
3-fluoro-4-chloro phenol, 3-fluoro-4-chloro-phenol, 3-fluoro-4-chlorophenol, 3-fluoro-4-chlorophenol;, 4-chlor-3-fluor-phenol;, 4-chloro-3-fluoro-phenol, 4-chloro-3-fluorophenol;, acmc-1ctvv, phenol, 4-chloro-3-fluoro, pubchem1493
OC1=CC=C(Cl)C(F)=C1
146.55
146.55
≥97.5% (GC)
Authentic
Colorless to Orange
5g
348-60-7
100.0
C6H4ClFO
MFCD00042583
XLHYAEBESNFTCA-UHFFFAOYSA-N
4-chloro-3-fluorophenol
2724523
Liquid

Chemical Identifiers

348-60-7
146.55
XLHYAEBESNFTCA-UHFFFAOYSA-N
2724523
OC1=CC=C(Cl)C(F)=C1
C6H4ClFO
MFCD00042583
3-fluoro-4-chloro phenol, 3-fluoro-4-chloro-phenol, 3-fluoro-4-chlorophenol, 3-fluoro-4-chlorophenol;, 4-chlor-3-fluor-phenol;, 4-chloro-3-fluoro-phenol, 4-chloro-3-fluorophenol;, acmc-1ctvv, phenol, 4-chloro-3-fluoro, pubchem1493
4-chloro-3-fluorophenol
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents