missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Chloro-3-fluorophenol, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 249970010

 View more versions of this product

Catalog No. AC249970010

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

4-Chloro-3-fluorophenol
Colorless to pale yellow, may discolor to orange during storage
1g
348-60-7
Glass bottle
Liquid
98%
OHC6H3ClF
MFCD00042583
Authentic
50° to 55°C
98%
97.5
100.0
≥97.5% (GC)
C6H4ClFO
146.55

Chemical Identifiers

348-60-7
100.0
C6H4ClFO
146.55
4-Chloro-3-fluorophenol, 98%
97.5
98%
OHC6H3ClF
MFCD00042583
≥97.5% (GC)

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning