Please login to your online account to display your discounted pricing

4-Chloro-3-fluorophenol, 98%, ACROS Organics™

$93.96 - $93.96

Chemical Identifiers

Chemical Name or Material 4-Chloro-3-fluorophenol
Name Note 98%
CAS 348-60-7
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5% (GC)
Assay Percent Range 98%
Molecular Formula C6H4ClFO
Linear Formula OHC6H3ClF
Formula Weight 146.55
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC249970050

  SDS
thermo fisher scientific
249970050
5g Each for $93.96

AC249970010

  SDS
thermo fisher scientific
249970010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5% (GC)
Physical Form Liquid
Formula Weight 146.55
Quantity 5g
Color Colorless to pale yellow, may discolor to orange during storage
Melting Point 50° to 55°C
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C6H4ClFO
Linear Formula OHC6H3ClF
Health Hazard 2 GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Chemical Name or Material 4-Chloro-3-fluorophenol
Infrared Spectrum Authentic
Name Note 98%
MDL Number MFCD00042583
CAS 348-60-7
CAS Min % 97.5
Health Hazard 1 GHS Signal Word: Warning
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.