missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-3-fluorobenzeneboronic acid pinacol ester, 96%

Manufacturer:  Alfa Aesar™ H6244503

 View more versions of this product

Catalog No. AAH6244503



Specifications

Specifications

4-Chloro-3-fluorobenzeneboronic acid pinacol ester
627525-83-1
MFCD16996258
XJZYKURDYQAGGN-UHFFFAOYSA-N
2-(4-chloro-3-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
22240146
Solid
1g
C12H15BClFO2
1,3,2-dioxaborolane, 2-4-chloro-3-fluorophenyl-4,4,5,5-tetramethyl
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)Cl)F
256.508
256.51
96%

Chemical Identifiers

627525-83-1
256.508
XJZYKURDYQAGGN-UHFFFAOYSA-N
22240146
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)Cl)F
C12H15BClFO2
MFCD16996258
1,3,2-dioxaborolane, 2-4-chloro-3-fluorophenyl-4,4,5,5-tetramethyl
2-(4-chloro-3-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

Documents