missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-3-fluorobenzeneboronic acid pinacol ester, 96%

$220.66 - $663.68

Chemical Identifiers

CAS 627525-83-1
Molecular Formula C12H15BClFO2
Molecular Weight (g/mol) 256.508
MDL Number MFCD16996258
InChI Key XJZYKURDYQAGGN-UHFFFAOYSA-N
Synonym 1,3,2-dioxaborolane, 2-4-chloro-3-fluorophenyl-4,4,5,5-tetramethyl, 2-4-chloro-3-fluoro-phenyl-4,4,5,5-tetramethyl-1,3,2 dioxaborolane, 2-4-chloro-3-fluorophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-chloro-3-fluorobenzeneboronic acid pinacol ester, 4-chloro-3-fluorophenylboronic acid pinacol ester
PubChem CID 22240146
IUPAC Name 2-(4-chloro-3-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)Cl)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH62445MD View Documents Alfa Aesar™
H62445MD
250mg Each for $220.66
 
AAH6244503 View Documents Alfa Aesar™
H6244503
1g Each for $663.68
 
Specifications

Chemical Identifiers

627525-83-1
256.508
XJZYKURDYQAGGN-UHFFFAOYSA-N
22240146
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)Cl)F
C12H15BClFO2
MFCD16996258
1,3,2-dioxaborolane, 2-4-chloro-3-fluorophenyl-4,4,5,5-tetramethyl, 2-4-chloro-3-fluoro-phenyl-4,4,5,5-tetramethyl-1,3,2 dioxaborolane, 2-4-chloro-3-fluorophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-chloro-3-fluorobenzeneboronic acid pinacol ester, 4-chloro-3-fluorophenylboronic acid pinacol ester
2-(4-chloro-3-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Specifications

250mg
C12H15BClFO2
1,3,2-dioxaborolane, 2-4-chloro-3-fluorophenyl-4,4,5,5-tetramethyl, 2-4-chloro-3-fluoro-phenyl-4,4,5,5-tetramethyl-1,3,2 dioxaborolane, 2-4-chloro-3-fluorophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-chloro-3-fluorobenzeneboronic acid pinacol ester, 4-chloro-3-fluorophenylboronic acid pinacol ester
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)Cl)F
256.508
256.51
96%
627525-83-1
MFCD16996258
XJZYKURDYQAGGN-UHFFFAOYSA-N
2-(4-chloro-3-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
22240146
Solid
4-Chloro-3-fluorobenzeneboronic acid pinacol ester
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures